Learn More
Medchemexpress LLC Tezacaftor | 1152311-62-0 | C26H27F3N2O6 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY15448100MG
Encompass_Preferred
Tezacaftor is a cystic fibrosis transmembrane conductance regulator (CFTR) corrector that assists CFTR protein in reaching the cell surface. When combined with a CFTR potentiator, it demonstrates strong effectiveness against cystic fibrosis and diseases involving a homozygous CFTR mutation. Another corrector, when combined with tezacaftor and a potentiator, targets cystic fibrosis with at least one specific mutation, often helping to avoid the need for lung transplantation.
  • Helps protein reach the cell surface.
  • Increases protein expression levels in specific cells.
  • Shows potent efficacy against certain genetic conditions when combined with other compounds.
  • Can improve clinical outcomes in patients with specific mutations.
  • Aids in prolonging the opening time of cell surface protein channels when used with a potentiator.
Catalog No. 50-003-76272