Learn More
Medchemexpress LLC HY-15448 50mg , Tezacaftor CAS:1152311-62-0 Purity:>98%
SDP
Catalog No. 502260911
Encompass_Preferred
HY-15448 50mg Tezacaftor CAS: 1152311-62-0 Tezacaftor (VX-661) is a second F508del CFTR corrector and help CFTR protein reach the cell surface. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-226-0911 Supplier Medchemexpress LLC Supplier No. HY1544850MG
HY-15448 50mg Tezacaftor CAS: 1152311-62-0 Tezacaftor (VX-661) is a second F508del CFTR corrector and help CFTR protein reach the cell surface. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.