Learn More
Selleck Chemical LLC Tezacaftor
SDP
Catalog No. 5000002148
Encompass_Preferred
Tezacaftor (VX-661) is a second F508del CFTR corrector and is believed to help CFTR protein reach the cell surface Phase 2
Catalog No. 50-000-02148 Supplier Selleck Chemical LLC Supplier No. S705950MG
Tezacaftor (VX-661) is a second F508del CFTR corrector and is believed to help CFTR protein reach the cell surface Phase 2