Learn More
Medchemexpress LLC Tezacaftor (racemate) | 1226709-85-8 | 96.3% | 520.50 | C26H27F3N2O6 | 10 MG
SDP
Catalog No. 5000286627
Encompass_Preferred
(Rac)-Tezacaftor is the racemic form of tezacaftor, a small-molecule CFTR corrector used in research on F508del-associated cystic fibrosis. It is intended for in vitro cellular and biochemical studies of CFTR folding, trafficking, and pharmacological rescue, and is supplied as a high-purity research reagent suitable for assay development and compound handling workflows.
  • Racemic form of tezacaftor suitable for research.
  • Acts as a F508del CFTR corrector in cellular assays.
  • High purity for reproducible experimental results.
  • Available in small pre-weighed quantities for assay use.
  • Molecular formula and weight provided for formulation and calculations.
Catalog No. 50-002-86627 Supplier Medchemexpress LLC Supplier No. HY15448A10MG
(Rac)-Tezacaftor is the racemic form of tezacaftor, a small-molecule CFTR corrector used in research on F508del-associated cystic fibrosis. It is intended for in vitro cellular and biochemical studies of CFTR folding, trafficking, and pharmacological rescue, and is supplied as a high-purity research reagent suitable for assay development and compound handling workflows.
  • Racemic form of tezacaftor suitable for research.
  • Acts as a F508del CFTR corrector in cellular assays.
  • High purity for reproducible experimental results.
  • Available in small pre-weighed quantities for assay use.
  • Molecular formula and weight provided for formulation and calculations.