Learn More
Cayman Chemical TF CREB (Phospho-Ser133) Primary Antibody, 1 ea
SDP
Catalog No. 502886908
Encompass_Preferred
For laboratory research use only. This reagent is not for human or veterinary use. This product has been tested and formulated to work exclusively with the CREB (Phospho-Ser133) Transcription Factor Assay Kit from Cayman (Item No. 10009846). This product may not perform as described if used with other assay reagents or protocols.
Catalog No. 50-288-6908 Supplier Cayman Chemical Supplier No. 100098481 EA
For laboratory research use only. This reagent is not for human or veterinary use. This product has been tested and formulated to work exclusively with the CREB (Phospho-Ser133) Transcription Factor Assay Kit from Cayman (Item No. 10009846). This product may not perform as described if used with other assay reagents or protocols.