Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STA Pharmaceutical TFA C6 amine amidite | 1 g | CAS 133975-85-6 | MDL MFCD00797447

Catalog No. 502357191
Encompass

TFA C6 amine amidite is a Amidite reagent (Subcategory: Amino Modifiers) sold by WuXi TIDES. Offered in 1 g. Store at -20 °C. Ships on dry ice. SDS available for reference.

Specifications
- CAS: 133975-85-6
- MDL: MFCD00797447
- InChIKey: SMMKTKILBQHSFL-UHFFFAOYSA-N
- Molecular Weight: 413.421971487
- Molecular Formula: C17H31F3N3O3P
- Purity: ≥95%
- Container Type: 60 mL Glass (Septum)
- Pack Size: 1 g
- Net Weight: 1 g
- Gross Weight: 38.3 g
- Commodity Code: 29299090
- Country Of Origin: China
- IUPAC: 2-cyanoethyl (6-(2,2,2-trifluoroacetamido)hexyl) diisopropylphosphoramidite
- SMILES: CC(N(P(OCCCCCCNC(C(F)(F)F)=O)OCCC#N)C(C)C)C

Catalog No. 50-235-7191 Supplier STA Pharmaceutical Supplier No. STAAM10040AK601GM
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

TFA C6 amine amidite is a Amidite reagent (Subcategory: Amino Modifiers) sold by WuXi TIDES. Offered in 1 g. Store at -20 °C. Ships on dry ice. SDS available for reference.

Specifications
- CAS: 133975-85-6
- MDL: MFCD00797447
- InChIKey: SMMKTKILBQHSFL-UHFFFAOYSA-N
- Molecular Weight: 413.421971487
- Molecular Formula: C17H31F3N3O3P
- Purity: ≥95%
- Container Type: 60 mL Glass (Septum)
- Pack Size: 1 g
- Net Weight: 1 g
- Gross Weight: 38.3 g
- Commodity Code: 29299090
- Country Of Origin: China
- IUPAC: 2-cyanoethyl (6-(2,2,2-trifluoroacetamido)hexyl) diisopropylphosphoramidite
- SMILES: CC(N(P(OCCCCCCNC(C(F)(F)F)=O)OCCC#N)C(C)C)C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.