Learn More
Waters Corp Waters TFA Mixer Assembly, 380 μL, Zirconia, 15K psi
Catalog No. 502039233 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This TFA Mixer Assembly, featuring a 380 μL volume and zirconia construction, is designed as an optional component for ACQUITY UPLC Binary Solvent Manager systems. It is rated for high-pressure applications, up to 15,000 psi, ensuring reliable performance in demanding laboratory environments.
  • Designed for ACQUITY UPLC Binary Solvent Manager systems.
  • Features a 380 μL mixing volume.
  • Constructed with durable zirconia material.
  • Capable of withstanding pressures up to 15,000 psi (1034 bar).
  • Includes components for installation.
Catalog No. 50-203-9233 Supplier Waters Corp Supplier No. 205000705
This TFA Mixer Assembly, featuring a 380 μL volume and zirconia construction, is designed as an optional component for ACQUITY UPLC Binary Solvent Manager systems. It is rated for high-pressure applications, up to 15,000 psi, ensuring reliable performance in demanding laboratory environments.
  • Designed for ACQUITY UPLC Binary Solvent Manager systems.
  • Features a 380 μL mixing volume.
  • Constructed with durable zirconia material.
  • Capable of withstanding pressures up to 15,000 psi (1034 bar).
  • Includes components for installation.