Learn More
Aviva Systems Biology Corp TFAM Antibody - N-terminal region (ARP31400_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31400_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Transcription Factor AntibodiesAM. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TFAM. Alternate names: TCF6, MtTF1, mtTFA, TCF6L1, TCF6L2, TCF6L3ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit
Catalog No. 50-014-54207