Learn More
Aviva Systems Biology Corp Tfam Antibody - C-terminal region (ARP36993_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36993_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Tfam. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of mouse Tfam. Alternate names: AI661103, Hmgts, mtTFA, tsHMGApplicationIHC, WB. Species reactivity: Cow, Mouse, Rabbit, Rat
Catalog No. 50-014-56304