Learn More
Aviva Systems Biology Corp TFAM Antibody - middle region (ARP36992_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP36992_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TFAM. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of mouse TFAM. Alternate names: Hmgts, mtTFA, tsHMG, AI661103ApplicationWB. Species reactivity: Mouse, Rabbit, Rat
Catalog No. 50-014-56303