Learn More
ABclonal Technology TFAM Rabbit pAb
SDP
Supplier:  ABclonal Technology A135521000UL
Encompass_Preferred
This gene encodes a key mitochondrial transcription factor containing two high mobility group motifs. The encoded protein also functions in mitochondrial DNA replication and repair. Sequence polymorphisms in this gene are associated with Alzheimers and Parkinsons diseases. There are pseudogenes for this gene on chromosomes 6, 7, and 11. Alternative splicing results in multiple transcript variants.
Catalog No. 50-001-15608