Learn More
Medchemexpress LLC HY-112731 5mg Medchemexpress, TFAP CAS:1011244-68-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1127315MG
Encompass_Preferred
Medchemexpress, HY-112731 5mg TFAP CAS:1011244-68-0 TFAP is a selective cyclooxygenase-1 (COX-1) inhibitor, with an IC50 of 0.8 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9161