Learn More
Aviva Systems Biology Corp TFAP2B Antibody - N-terminal region (ARP38282_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38282_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TFAP2B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TFAP2B. Alternate names: AP-2B, AP2-B, MGC21381ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat, Zebrafish
Catalog No. 50-014-56593