Learn More
Aviva Systems Biology Corp TFAP2C Antibody - N-terminal region (ARP38284_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38284_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against TFAP2C. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human TFAP2C. Alternate names: AP2-GAMMA, ERF1, TFAP2G, hAP-2gApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-56595