Learn More
Aviva Systems Biology Corp TFAP2D Antibody - C-terminal region (ARP30084_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP30084_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against TFAP2D. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human TFAP2D. Alternate names: TFAP2BL1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-54119