Learn More
ProSci TFAP2D Peptide

Description
Specifications
Specifications
| Gene ID (Entrez) | 83741 |
| Accession Number | NP_758438.2 |
| Common Name | transcription factor AP-2 delta (activating enhancer binding protein 2 delta) |
| Content And Storage | Store at -20°C. When reconstituted minimize freezing and thawing. |
| Format | Lyophilized |
| Formulation | TFAP2D peptide is supplied as 100 μg of dried peptide. When the peptide is reconstituted in 200 μL water, the concentration will be 0.5 mg/mL |
| For Use With (Application) | Blocking Assay |
| Gene Symbol | TFAP2D |
| Quantity | 0.1 mg |
| Sequence | TFAP2D peptide corresponds to 14 amino acids near an internal region of TFAP2D. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.