Learn More
Aviva Systems Biology Corp TFAP2E Antibody - C-terminal region (ARP40023_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40023_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against TFAP2E. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human TFAP2E. Alternate names: AP2E, MGC49007ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53169