Learn More
Aviva Systems Biology Corp TFAP2E Antibody - N-terminal region (ARP31976_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31976_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TFAP2E. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TFAP2E. Alternate names: AP2E, MGC49007ApplicationWB. Species reactivity: Cow, Human, Mouse
Catalog No. 50-014-54398