Learn More
Aviva Systems Biology Corp TFAP4 Antibody - C-terminal region (ARP33435_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33435_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against TFAP4. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the C terminal region of human TFAP4. Alternate names: AP-4, bHLHc41ApplicationIHC, WB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55023