Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tfb

Catalog No. 901101250MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-110-1250MG 250 mg
90-110-11G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 90-110-1250MG Supplier Sigma Aldrich Supplier No. 901101250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

TFB, a hole transporting material and an electron-blocking layer, has high hole mobility, low electron affinity, and high ionic potential. Its electron blocking nature results in effective confinement of injected charge carriers in the perovskite layers.TFB can be used in the formation of multilayer quantum dot-based light-emitting diodes (LEDs). It can also be used in the fabrication of highly responsive gas sensors for breath analysis.

Specifications

CAS 220797-16-0
Linear Formula (C51H61N)n
Quantity 250 mg
Synonym Poly[(9,9-dioctylfluorenyl-2,7-diyl)-co-(4,4 -(N-(4-sec-butylphenyl)diphenylamine)]
Molecular Weight (g/mol) 30000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.