Learn More
Cayman Chemical TFB-TBOA (hydrate), Purity: ≥98%, 10 mg
SDP
Catalog No. 502990815
Encompass_Preferred
An inhibitor of EAAT1 and EAAT2 (IC50s = 22 and 17 nM for EAAT1 and EAAT2, respectively); selective for EAAT1 and EAAT2 over EAAT3 (IC50 = 300 nM), as well as a variety of neurotransmitter transporters and receptors at 10 µM; inhibits L-glutamate- and L-aspartate-induced currents in X. laevis oocytes at 10 µM; induces convulsions in mice at 0.2 µg/animal
Catalog No. 50-299-0815 Supplier Cayman Chemical Supplier No. 3141710 MG
An inhibitor of EAAT1 and EAAT2 (IC50s = 22 and 17 nM for EAAT1 and EAAT2, respectively); selective for EAAT1 and EAAT2 over EAAT3 (IC50 = 300 nM), as well as a variety of neurotransmitter transporters and receptors at 10 µM; inhibits L-glutamate- and L-aspartate-induced currents in X. laevis oocytes at 10 µM; induces convulsions in mice at 0.2 µg/animal