Learn More
Aviva Systems Biology Corp TFB1M Antibody - middle region (ARP34544_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP34544_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against TFB1M. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TFB1M. Alternate names: CGI-75, CGI75, mtTFB, mtTFB1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-55442