Learn More
Aviva Systems Biology Corp TFB2M Antibody - N-terminal region (ARP39549_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39549_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TFB2M. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TFB2M. Alternate names: FLJ22661, FLJ23182, Hkp1, mtTFB2ApplicationWB. Species reactivity: Human
Catalog No. 50-014-62878