Learn More
Aviva Systems Biology Corp TFCP2L1 Antibody - N-terminal region (ARP31918_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP31918_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TFCP2L1. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human TFCP2L1. Alternate names: CRTR1, LBP-9, LBP9ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast
Catalog No. 50-014-54372