Learn More
Aviva Systems Biology Corp TFDP1 Antibody - middle region (P100879_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp P100879_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TFDP1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TFDP1. Alternate names: DP1, DRTF1, Dp-1ApplicationWB. Species reactivity: Cow, Human, Mouse, Zebrafish
Catalog No. 50-014-62759