Learn More
Aviva Systems Biology Corp Tfdp2 Antibody - C-terminal region (ARP37585_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP37585_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Tfdp2. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: A330080J22Rik, DP-3, DP3, 1110029I05RikApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-284-661