Learn More
Aviva Systems Biology Corp TFDP3 Antibody - middle region (ARP58171_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58171_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TFDP3. It was validated on Western Blot. TFDP3,transcription factor Dp family, member 3,DP4; CT30; HCA661; MGC161639; TFDP3,transcription factor Dp family member 3,transcription factor Dp family member 3,E2F-like protein,OTTHUMP00000024051,cancer/testis antigen 30,hepatocellular carcinoma-associated antigen 661The immunogen is a synthetic peptide directed towards the middle region of human TFDP3. Alternate names: CT30, E2F-like, HCA661, MGC161639, DP4ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-286-448