Learn More
Aviva Systems Biology Corp TFDP3 Antibody - middle region (ARP32159_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32159_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TFDP3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TFDP3. Alternate names: CT30, E2F-like, HCA661, MGC161639, DP4ApplicationWB. Species reactivity: Human
Catalog No. 50-014-53860