Learn More
Ace Glass, Inc. PTFE stir blade, oval, 19x40mm, 10mm shafts
SDP
Catalog No. 509558236
Encompass_Preferred
19x40mm oval PTFE stir blade for 10mm O.D. button-type stir shafts.
Catalog No. 50-955-8236 Supplier Ace Glass, Inc. Supplier No. 808202
19x40mm oval PTFE stir blade for 10mm O.D. button-type stir shafts.