Learn More
TARGETMOL CHEMICALS INC TFEB ACTIVATOR 1 25MG
SDP
Catalog No. 502579104
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. TFEB activator 1 (Curcumin analog compound C1) is an activator of transcription factor EB (TFEB) with promise for the prevention or treatment of Alzheimer's disease. purity: 99%
Catalog No. 50-257-9104 Supplier TARGETMOL CHEMICALS INC Supplier No. T805525MG
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. TFEB activator 1 (Curcumin analog compound C1) is an activator of transcription factor EB (TFEB) with promise for the prevention or treatment of Alzheimer's disease. purity: 99%