Learn More
Medchemexpress LLC TFEB Decoy ODN sodium | 90.2% | 1 MG
SDP
Catalog No. 5000359737
Encompass_Preferred
TFEB Decoy ODN sodium is a synthetic oligonucleotide featuring a hairpin ring structure, engineered to inhibit Transcription factor EB (TFEB). It has demonstrated efficacy in inhibiting fibrosis and autophagy within a UUO mouse model, and also exhibits anti-inflammatory effects.
  • Synthetic oligonucleotide with a hairpin ring structure.
  • Designed to inhibit transcription factor EB (TFEB).
  • Inhibited fibrosis in a UUO mouse model.
  • Inhibited autophagy in a UUO mouse model.
  • Showed anti-inflammatory effects.
Catalog No. 50-003-59737 Supplier Medchemexpress LLC Supplier No. HY1482301MG
TFEB Decoy ODN sodium is a synthetic oligonucleotide featuring a hairpin ring structure, engineered to inhibit Transcription factor EB (TFEB). It has demonstrated efficacy in inhibiting fibrosis and autophagy within a UUO mouse model, and also exhibits anti-inflammatory effects.
  • Synthetic oligonucleotide with a hairpin ring structure.
  • Designed to inhibit transcription factor EB (TFEB).
  • Inhibited fibrosis in a UUO mouse model.
  • Inhibited autophagy in a UUO mouse model.
  • Showed anti-inflammatory effects.