Learn More
Medchemexpress LLC TFEB Decoy ODN sodium | 90.2% | 5 MG
SDP
Catalog No. 5000359775
Encompass_Preferred
TFEB Decoy ODN sodium is a synthetic oligonucleotide designed with a hairpin ring structure to inhibit Transcription factor EB (TFEB).
  • Inhibits fibrosis
  • Inhibits autophagy in a UUO mouse model
  • Demonstrates anti-inflammatory effects
Catalog No. 50-003-59775 Supplier Medchemexpress LLC Supplier No. HY1482305MG
TFEB Decoy ODN sodium is a synthetic oligonucleotide designed with a hairpin ring structure to inhibit Transcription factor EB (TFEB).
  • Inhibits fibrosis
  • Inhibits autophagy in a UUO mouse model
  • Demonstrates anti-inflammatory effects