Learn More
Cell Signaling Technology TFEB Signaling Antibody Sampler Kit, 1 Kit
SDP
Catalog No. 502732399
Encompass
The TFEB Signaling Antibody Sampler Kit provides an economical means of analyzing the regulation of transcription factor EB (TFEB). This kit contains an array of primary antibodies designed to detect TFEB and its phosphorylation states, as well as key upstream regulators including mTOR and calcineurin, enabling comprehensive study of the autophagy signaling pathway.
  • Includes antibodies for TFEB, Phospho-TFEB (Ser211), Phospho-TFEB (Ser122), mTOR, Phospho-mTOR (Ser2448), and pan-calcineurin A
  • Supplied with HRP-linked secondary antibody for convenient western blot detection
  • Optimized for the analysis of signaling pathways involved in lysosome biogenesis and autophagy
  • Validated for multiple research applications
  • Sufficient antibody quantity for two western blot experiments per primary antibody
Catalog No. 50-273-2399 Supplier Cell Signaling Technology Supplier No. 63734T
The TFEB Signaling Antibody Sampler Kit provides an economical means of analyzing the regulation of transcription factor EB (TFEB). This kit contains an array of primary antibodies designed to detect TFEB and its phosphorylation states, as well as key upstream regulators including mTOR and calcineurin, enabling comprehensive study of the autophagy signaling pathway.
  • Includes antibodies for TFEB, Phospho-TFEB (Ser211), Phospho-TFEB (Ser122), mTOR, Phospho-mTOR (Ser2448), and pan-calcineurin A
  • Supplied with HRP-linked secondary antibody for convenient western blot detection
  • Optimized for the analysis of signaling pathways involved in lysosome biogenesis and autophagy
  • Validated for multiple research applications
  • Sufficient antibody quantity for two western blot experiments per primary antibody