Learn More
Aviva Systems Biology Corp TFEC Antibody - N-terminal region (ARP32279_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32279_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TFEC. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TFEC. Alternate names: TCFEC, TFECL, TFE-C, TFEC-L, bHLHe34, hTFEC-LApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-54508