Learn More
Cayman Chemical TFF1 trefoil factor 1 or pS2
SDP
Catalog No. 502948342
Encompass_Preferred
An alkaloid inhibits the growth of human A375 A431 A549 MES-SA and MCF-7 cells (IC50 3 nM for all) inhibits hypoxia-induced gene expression in an HRE reporter assay (IC50 8.7 nM) prevents lesion formation in TMV-infected tobacco plants at 500 Ug/ml inhibits protein synthesis by yeast and mammalian ribosomes
Catalog No. 50-294-8342 Supplier Cayman Chemical Supplier No. 33081100 UL
An alkaloid inhibits the growth of human A375 A431 A549 MES-SA and MCF-7 cells (IC50 3 nM for all) inhibits hypoxia-induced gene expression in an HRE reporter assay (IC50 8.7 nM) prevents lesion formation in TMV-infected tobacco plants at 500 Ug/ml inhibits protein synthesis by yeast and mammalian ribosomes