Learn More
BioVendor, LLC Trefoil Factor 3, Polyclonal Antibody ELISA
SDP
Supplier:  BioVendor, LLC RD184160100
Encompass
Trefoil Factor 3, TypePolyclonal AntibodyApplicationsWestern blotting, ELISA, Immunohistochemistry Source of AntigenE. coliHostsSheepPreparationThe antibody was raised in sheep by immunization with the recombinant Human Trefoil Factor 3.Amino Acid SequenceThe immunization antigen (7.82 kDa ¿ calculated) is a protein containing 69 AA of recombinant Human Trefoil Factor 3. N-terminal His-tag, 10 extra AA (highlighted).MKHHHHHHASEEYVGLSANQCAVPAKDRVDCGYPHVTPKECNNRGCCFDSRIPGVPWCFKPLQEAECTFSpecies ReactivityHuman. Not yet tested in other species.Purification MethodImmunoaffinity chromatography on a column with immobilized recombinant Human Trefoil Factor 3.Antibody Content0.1 mg (determined by BCA method, BSA was used as a standard)FormulationThe antibody is lyophilized in 0.05 M phosphate buffer, 0.1 M NaCl, pH 7.2. AZIDE FREE.
Catalog No. 50-379-7