Learn More
Medchemexpress LLC TFLLR-NH2(TFA) | 1313730-19-6 | 99.6% | 761.83 | 10 MG
SDP
Catalog No. 5000425648
Encompass_Preferred
TFLLR-NH2(TFA) is a selective PAR1 agonist with an EC50 of 1.9 μM, intended for research use. It stimulates concentration-dependent increases in intracellular calcium and promotes upregulation of E-cadherin expression in SW620 cells. In vivo, it can induce oedema and extravasation in various organs.
  • Selective PAR1 agonist (EC50 of 1.9 μM)
  • Induces concentration-dependent increases in intracellular calcium
  • Upregulates E-cadherin expression in SW620 cells
  • Stimulates secreted TGF-β1 in platelet cultures
  • Can induce oedema and extravasation in animal models
  • Stable for 2 years as powder at -80°C
  • Stable for 6 months in solvent at -80°C
Catalog No. 50-004-25648 Supplier Medchemexpress LLC Supplier No. HYP0226A10MG
TFLLR-NH2(TFA) is a selective PAR1 agonist with an EC50 of 1.9 μM, intended for research use. It stimulates concentration-dependent increases in intracellular calcium and promotes upregulation of E-cadherin expression in SW620 cells. In vivo, it can induce oedema and extravasation in various organs.
  • Selective PAR1 agonist (EC50 of 1.9 μM)
  • Induces concentration-dependent increases in intracellular calcium
  • Upregulates E-cadherin expression in SW620 cells
  • Stimulates secreted TGF-β1 in platelet cultures
  • Can induce oedema and extravasation in animal models
  • Stable for 2 years as powder at -80°C
  • Stable for 6 months in solvent at -80°C