Learn More
Medchemexpress LLC TFLLR-NH2(TFA) | 1313730-19-6 | 99.6% | 761.83 | 25 MG
SDP
Catalog No. 5000426820
Encompass_Preferred
TFLLR-NH2 (TFA) is a selective PAR1 agonist with an EC50 of 1.9 μM. It stimulates concentration-dependent increases in [Ca2+]i, upregulates E-cadherin, and downregulates vimentin expression in SW620 cells. It also induces a dose-dependent increase of secreted TGF-β1 in activated platelets. In vivo, it causes marked and sustained oedema and Evans blue extravasation, and produces contraction and relaxation in rat duodenal smooth muscle.
  • Selective PAR1 agonist with an EC50 of 1.9 μM
  • Appears as a white to off-white solid
  • Soluble in DMSO and H2O at 100 mg/mL
  • Upregulates E-cadherin and downregulates vimentin expression
  • Induces dose-dependent increase of secreted TGF-β1
  • Stimulates oedema and Evans blue extravasation in vivo
  • Produces contraction and relaxation in rat duodenal smooth muscle
Catalog No. 50-004-26820 Supplier Medchemexpress LLC Supplier No. 25MGHYP0226A
TFLLR-NH2 (TFA) is a selective PAR1 agonist with an EC50 of 1.9 μM. It stimulates concentration-dependent increases in [Ca2+]i, upregulates E-cadherin, and downregulates vimentin expression in SW620 cells. It also induces a dose-dependent increase of secreted TGF-β1 in activated platelets. In vivo, it causes marked and sustained oedema and Evans blue extravasation, and produces contraction and relaxation in rat duodenal smooth muscle.
  • Selective PAR1 agonist with an EC50 of 1.9 μM
  • Appears as a white to off-white solid
  • Soluble in DMSO and H2O at 100 mg/mL
  • Upregulates E-cadherin and downregulates vimentin expression
  • Induces dose-dependent increase of secreted TGF-β1
  • Stimulates oedema and Evans blue extravasation in vivo
  • Produces contraction and relaxation in rat duodenal smooth muscle