Learn More
Medchemexpress LLC Tfllr-nh2(tfa) | 1313730-19-6 | 99.6% | 761.83 | 5 MG
SDP
Catalog No. 5000425817
Encompass_Preferred
TFLLR-NH2 (TFA) is a selective PAR1 agonist with an EC50 of 1.9 μM, intended for research use only. It appears as a white to off-white solid.
  • Acts as a selective PAR1 agonist
  • Stimulates concentration-dependent increases in [Ca2+]i in neurons
  • Activated platelets upregulate E-cadherin and downregulate vimentin expression in SW620 cells
  • Increases secreted TGF-β1 in platelet cultures
  • Injection into rat paws stimulates marked and sustained oedema
  • Stimulates Evans blue extravasation in various organs in wild-type mice
  • Can induce both contraction and relaxation in rat duodenal smooth muscle
  • Recommended storage for powder is -80°C for 2 years or -20°C for 1 year
  • Recommended storage in solvent is -80°C for 6 months or -20°C for 1 month
  • High solubility in DMSO and H2O (100 mg/mL)
Catalog No. 50-004-25817 Supplier Medchemexpress LLC Supplier No. HYP0226A5MG
TFLLR-NH2 (TFA) is a selective PAR1 agonist with an EC50 of 1.9 μM, intended for research use only. It appears as a white to off-white solid.
  • Acts as a selective PAR1 agonist
  • Stimulates concentration-dependent increases in [Ca2+]i in neurons
  • Activated platelets upregulate E-cadherin and downregulate vimentin expression in SW620 cells
  • Increases secreted TGF-β1 in platelet cultures
  • Injection into rat paws stimulates marked and sustained oedema
  • Stimulates Evans blue extravasation in various organs in wild-type mice
  • Can induce both contraction and relaxation in rat duodenal smooth muscle
  • Recommended storage for powder is -80°C for 2 years or -20°C for 1 year
  • Recommended storage in solvent is -80°C for 6 months or -20°C for 1 month
  • High solubility in DMSO and H2O (100 mg/mL)