Learn More
Medchemexpress LLC HY-P0226A 1mg Medchemexpress, TFLLR-NH2(TFA) CAS:1313730-19-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYP0226A1MG
Encompass_Preferred
Medchemexpress, HY-P0226A 1mg TFLLR-NH2(TFA) CAS:1313730-19-6 TFLLR-NH2 (TFA) is a selective PAR1 agonist with an EC 50 of 1.9 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-4060