Learn More
Perkin Elmer US LLC TFM Cap for Standard 75 mL, 40 Bar Digestion Vessels without Pressure Sensor
Catalog No. 502111707
Encompass_Preferred
TFM Cap for Standard 75 mL (40 Bar) Digestion vessels without pressure sensor.
Catalog No. 50-211-1707 Supplier Perkin Elmer US LLC Supplier No. N3132012
TFM Cap for Standard 75 mL (40 Bar) Digestion vessels without pressure sensor.