Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Milestone Inc TFM Vessel, 900 ml, for Milestone microwave digestion systems
SDP

Catalog No. NC2272171
Encompass

The TFM vessel with a 900 ml capacity is engineered for microwave digestion systems, providing essential features for demanding laboratory applications. This high-quality vessel ensures robust chemical resistance and thermal stability, optimized for trace metal analysis and other sensitive procedures. The construction from TFM guarantees high purity, minimizing contamination risks, while its durable design supports safe and efficient digestion processes in various laboratory environments.

  • Capacity: 900 ml
  • Material: TFM
  • Application: Microwave digestion
  • Compatibility: Milestone microwave digestion systems
  • Chemical resistance: High
  • Thermal stability: High
  • Purity: High
  • Design: Durable for laboratory use

This item is not returnable. View return policy
Catalog No. NC2272171 Supplier Milestone Inc Supplier No. HB00070B
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The TFM vessel with a 900 ml capacity is engineered for microwave digestion systems, providing essential features for demanding laboratory applications. This high-quality vessel ensures robust chemical resistance and thermal stability, optimized for trace metal analysis and other sensitive procedures. The construction from TFM guarantees high purity, minimizing contamination risks, while its durable design supports safe and efficient digestion processes in various laboratory environments.

  • Capacity: 900 ml
  • Material: TFM
  • Application: Microwave digestion
  • Compatibility: Milestone microwave digestion systems
  • Chemical resistance: High
  • Thermal stability: High
  • Purity: High
  • Design: Durable for laboratory use

This item is not returnable. View return policy
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.