Learn More
Cayman Chemical TFMB-R-2-HG 10mg
SDP
Catalog No. 502984197
Encompass_Preferred
A cell-permeable form of (R)-2-HG; has been used to assess the role of (R)-2-HG in leukemogenesis
Catalog No. 50-298-4197 Supplier Cayman Chemical Supplier No. 2046110 MG
A cell-permeable form of (R)-2-HG; has been used to assess the role of (R)-2-HG in leukemogenesis