Learn More
Medchemexpress LLC TFMB-(R)-2-HG | 1445700-01-5 | 98.3% | 50 MG
SDP
Catalog No. 5000339531
Encompass_Preferred
TFMB-(R)-2-HG is a cell membrane-permeable (R)-2-HG and an acute myeloid leukemia (AML) oncogenic factor. It functions by competitively inhibiting alpha-ketoglutarate-dependent dioxygenases, such as KDM2B and FTO. This compound impairs cell differentiation when estrogen is withdrawn and is utilized in research concerning acute myeloid leukemia and glioma.
  • Cell membrane-permeable (R)-2-HG
  • Acute myeloid leukemia (AML) oncogenic factor
  • Competitively inhibits alpha-ketoglutarate-dependent dioxygenases (e.g., KDM2B, FTO)
  • Impairs cell differentiation upon estrogen withdrawal
  • Useful in acute myeloid leukemia research
  • Useful in glioma research
Catalog No. 50-003-39531 Supplier Medchemexpress LLC Supplier No. HY12907950MG
TFMB-(R)-2-HG is a cell membrane-permeable (R)-2-HG and an acute myeloid leukemia (AML) oncogenic factor. It functions by competitively inhibiting alpha-ketoglutarate-dependent dioxygenases, such as KDM2B and FTO. This compound impairs cell differentiation when estrogen is withdrawn and is utilized in research concerning acute myeloid leukemia and glioma.
  • Cell membrane-permeable (R)-2-HG
  • Acute myeloid leukemia (AML) oncogenic factor
  • Competitively inhibits alpha-ketoglutarate-dependent dioxygenases (e.g., KDM2B, FTO)
  • Impairs cell differentiation upon estrogen withdrawal
  • Useful in acute myeloid leukemia research
  • Useful in glioma research