Learn More
Medchemexpress LLC TFP-PEG3-TFP 50mg
SDP
Catalog No. 5000205719
Encompass_Preferred
TFP-PEG3-TFP is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs[1]
Catalog No. 50-002-05719 Supplier Medchemexpress LLC Supplier No. HYW09611250MG
TFP-PEG3-TFP is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs[1]