Learn More
Aviva Systems Biology Corp TFPI Antibody - C-terminal region (ARP63091_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63091_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TFPI. It was validated on Western Blot. . Alternate names: EPI, LACI, TFI, TFPI1ApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Pig, Rabbit, Rat
Catalog No. 50-287-871