Learn More
Medchemexpress LLC TFPI Human HEK293 10ug | 10ug
SDP
Catalog No. 5000186382
Encompass_Preferred
TFPI protein encoded by the TFPI gene is an important Kunitz-type serine protease inhibitor that regulates TF-dependent pathways in coagulation It inhibits factor X and VIIa-TF proteases preventing excessive clot formation TFPI Protein Human (HEK293 His) is the recombinant human-derived TFPI protein expressed by HEK293 with C-6 His labeled tag
Catalog No. 50-001-86382 Supplier Medchemexpress LLC Supplier No. HYP7722710UG
TFPI protein encoded by the TFPI gene is an important Kunitz-type serine protease inhibitor that regulates TF-dependent pathways in coagulation It inhibits factor X and VIIa-TF proteases preventing excessive clot formation TFPI Protein Human (HEK293 His) is the recombinant human-derived TFPI protein expressed by HEK293 with C-6 His labeled tag