Learn More
Medchemexpress LLC TFPI Mouse HEK293 10ug
SDP
Catalog No. 5000180858
Encompass_Preferred
TFPI Protein a potent inhibitor in the coagulation cascade directly inhibits factor X (Xa) In a Xa-dependent manner it also inhibits the activity of VIIa/tissue factor potentially forming a quaternary complex with Xa TFPI VIIa and tissue factor Beyond its antithrombotic role TFPI associates with plasma lipoproteins indicating its regulatory involvement in hemostasis and clotting processes TFPI Protein Mouse (HEK293 C-His) is the recombinant mouse-derived TFPI protein expressed by HEK293 with C-10 His labeled tag
Catalog No. 50-001-80858 Supplier Medchemexpress LLC Supplier No. HYP73425A10UG
TFPI Protein a potent inhibitor in the coagulation cascade directly inhibits factor X (Xa) In a Xa-dependent manner it also inhibits the activity of VIIa/tissue factor potentially forming a quaternary complex with Xa TFPI VIIa and tissue factor Beyond its antithrombotic role TFPI associates with plasma lipoproteins indicating its regulatory involvement in hemostasis and clotting processes TFPI Protein Mouse (HEK293 C-His) is the recombinant mouse-derived TFPI protein expressed by HEK293 with C-10 His labeled tag