Learn More
ACROBiosystems Recombinant Protein;Human TFPI-2 Protein;HEK293;50UG;TF2,TFPI2,PP5,REF1,TFPI-2,PP-5,REF-1
Catalog No. 501657816
Encompass_Preferred
Recombinant Protein;Human TFPI-2 Protein;50UG;Human TFPI-2, His Tag (TF2-H5227) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - Lys 213 (Accession # NP_006519.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 22.7 kDa. The protein migrates as 26-32 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-7816 Supplier ACROBiosystems Supplier No. TF2H522750UG
Recombinant Protein;Human TFPI-2 Protein;50UG;Human TFPI-2, His Tag (TF2-H5227) is expressed from human 293 cells (HEK293). It contains AA Asp 23 - Lys 213 (Accession # NP_006519.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 22.7 kDa. The protein migrates as 26-32 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.