Learn More
REPLIGEN CORPORATION Repligen ProConnex Standard Flow Path, 2L Capacity, Single-Use Assembly
SDP
Catalog No. NC2326874
Encompass
The Repligen ProConnex Standard Flow Path is a single-use assembly designed to enhance workflow efficiency in tangential flow filtration systems, particularly compatible with KrosFlo KR2i. It eliminates the need for cleaning and validation, allowing for reliable bioprocessing in manufacturing. The product features lot-traceable materials and compliance with rigorous industry standards, ensuring consistent performance in critical applications.
Catalog No. NC2326874 Supplier REPLIGEN CORPORATION Supplier No. STNGXGN15653S
The Repligen ProConnex Standard Flow Path is a single-use assembly designed to enhance workflow efficiency in tangential flow filtration systems, particularly compatible with KrosFlo KR2i. It eliminates the need for cleaning and validation, allowing for reliable bioprocessing in manufacturing. The product features lot-traceable materials and compliance with rigorous industry standards, ensuring consistent performance in critical applications.